missing translation for 'onlineSavingsMsg'
Learn More

(4S)-(3',4'-Dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one, TRC

Product Code. 30386311
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386311 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386311 Supplier Toronto Research Chemicals Supplier No. TRCD435706500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4S)-4-(3,4-dichlorophenyl)-3,4-dihydronaphthalen-1(2H)-one
CAS 124379-29-9
Molecular Formula C16 H12 Cl2 O
Quantity 500 mg
Synonym 1(2H)-Naphthalenone, 4-(3,4-dichlorophenyl)-3,4-dihydro-, (4S)-, 1(2H)-Naphthalenone, 4-(3,4-dichlorophenyl)-3,4-dihydro-, (S)-, (4S)-4-(3,4-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone, (4S)-(3,4-Dichlorophenyl)-3,4-dihydro-1(2H)-naphthalenone, (4S)-4-(3,4-Dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one, (4S)-4-(3,4-Dichlorophenyl)-3,4-dihydronaphthalen-1(2H)-one
InChI Formula InChI=1S/C16H12Cl2O/c17-14-7-5-10(9-15(14)18)11-6-8-16(19)13-4-2-1-3-12(11)13/h1-5,7,9,11H,6,8H2/t11-/m0/s1
SMILES Clc1ccc(cc1Cl)[C@@H]2CCC(=O)c3ccccc23
IUPAC Name (4S)-4-(3,4-dichlorophenyl)-3,4-dihydro-2H-naphthalen-1-one
Molecular Weight (g/mol) 291.1719
Formula Weight 290.0265
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.