missing translation for 'onlineSavingsMsg'
Learn More

(4S)-3-[(2R,5S)-5-(4-Fluorophenyl)-2-[(S)-[(3-fluorophenyl)amino][4-[(trimethylsilyl)oxy]phenyl]methyl]-1-oxo-5-[(trimethylsilyl)oxy]pentyl]-4-phenyl-2-oxazolidinone, TRC

Product Code. 30394476
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394476 250 mg 250mg
30393828 500 mg 500mg
30390053 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394476 Supplier Toronto Research Chemicals Supplier No. TRCF598460250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4S)-3-[(2R,5S)-5-(4-Fluorophenyl)-2-[(S)-[(3-fluorophenyl)amino][4-[(trimethylsilyl)oxy]phenyl]methyl]-1-oxo-5-[(trimethylsilyl)oxy]pentyl]-4-phenyl-2-oxazolidinone
Analyte or Component Names (4S)-3-[(2R,5S)-5-(4-Fluorophenyl)-2-[(S)-[(3-fluorophenyl)amino][4-[(trimethylsilyl)oxy]phenyl]methyl]-1-oxo-5-[(trimethylsilyl)oxy]pentyl]-4-phenyl-2-oxazolidinone
Shipping Condition Room Temperature
Molecular Formula C39 H46 F2 N2 O5 Si2
InChI Formula InChI=1S/C39H46F2N2O5Si2/c1-49(2,3)47-33-21-17-29(18-22-33)37(42-32-14-10-13-31(41)25-32)34(23-24-36(48-50(4,5)6)28-15-19-30(40)20-16-28)38(44)43-35(26-46-39(43)45)27-11-8-7-9-12-27/h7-22,25,34-37,42H,23-24,26H2,1-6H3/t34-,35+,36+,37-/m1/s1
Solution Type Neat
SMILES C[Si](C)(C)O[C@@H](CC[C@H]([C@H](Nc1cccc(F)c1)c2ccc(O[Si](C)(C)C)cc2)C(=O)N3[C@@H](COC3=O)c4ccccc4)c5ccc(F)cc5
IUPAC Name (4R)-3-[(2R,5S)-2-[(S)-(3-fluoroanilino)-(4-trimethylsilyloxyphenyl)methyl]-5-(4-fluorophenyl)-5-trimethylsilyloxypentanoyl]-4-phenyl-1,3-oxazolidin-2-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 716.961
Quantity 250 mg
Formula Weight 716.291
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.