missing translation for 'onlineSavingsMsg'
Learn More

(4S)-2-[Carboxy[(phenylacetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penicilloic Acids of Benzylpenicillin), Mikromol™

Product Code. 30049665
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049665 25 mg 25mg
17993936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30049665 Supplier Mikromol Supplier No. MM0160.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(4S)-2-[Carboxy[(phenylacetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penicilloic Acids of Benzylpenicillin)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4S)-2-[Carboxy[(phenylacetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penicilloic Acids of Benzylpenicillin)
CAS 13057-98-2
Analyte or Component Names (4S)-2-[Carboxy[(phenylacetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penicilloic Acids of Benzylpenicillin)
Shipping Condition Ice pack (-20°C)
Molecular Formula C16 H20 N2 O5 S
Certifications/Compliance ISO 9001
Synonym Benzylpenicillin Benzathine Imp. E (EP), Benzylpenicillin Potassium Imp. E (EP), Benzylpenicillin Procaine Imp. B (EP), (4S)-2-[Carboxy[(phenylacetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid, Penicilloic Acids of Benzylpenicillin, Benzylpenicillin Benzathine Impurity E, Benzylpenicillin Potassium Impurity E, Benzylpenicillin Procaine Impurity B, Carbenicillin Sodium Impurity G, Carbenicillin Impurity G, Benzylpenicillin Impurity E
InChI Formula InChI=1S/C16H20N2O5S/c1-16(2)12(15(22)23)18-13(24-16)11(14(20)21)17-10(19)8-9-6-4-3-5-7-9/h3-7,11-13,18H,8H2,1-2H3,(H,17,19)(H,20,21)(H,22,23)/t11?,12-,13?/m0/s1
SMILES CC1(C)SC(N[C@H]1C(=O)O)C(NC(=O)Cc2ccccc2)C(=O)O
IUPAC Name (4S)-2-[carboxy-[(2-phenylacetyl)amino]methyl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylic acid
Molecular Weight (g/mol) 352.41
Quantity 25 mg
Formula Weight 352.1093 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.