missing translation for 'onlineSavingsMsg'
Learn More

(4R)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid, 99%, Thermo Scientific™

Product Code. 10451012 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10451012 1 g 1g
10117983 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10451012 Supplier Thermo Scientific Acros Supplier No. 337840010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162856-35-1
Molecular Formula C10H19NO3Si
Molecular Weight (g/mol) 229.35
InChI Key LIEWITJXZYCDLE-SSDOTTSWSA-N
Synonym r-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid, 2r-1-tert-butyldimethylsilyl-4-oxoazetidine-2-carboxylic acid, 4r-n-tert-butyldimethylsilyl azetidin-2-one-carboxylic acid, 4r-n-tert-butyldimethylsilyl azetidin-2-one-4-carboxylic acid, 1-tert-butyldimethylsilyl-4-oxoazetidine-2 r-carboxylic acid, 2r-1-tert-butyl dimethyl silyl-4-oxoazetidine-2-carboxylic acid, r-1-tert-butyl-dimethyl-silanyl-4-oxo-azetidine-2-carboxylic acid, 2-azetidinecarboxylicacid, 1-1,1-dimethylethyl dimethylsilyl-4-oxo-, 2r, 2-azetidinecarboxylicacid,1-1,1-dimethylethyl dimethylsilyl-4-oxo-, 2r
PubChem CID 2733822
IUPAC Name (2R)-1-[tert-butyl(dimethyl)silyl]-4-oxoazetidine-2-carboxylic acid
SMILES CC(C)(C)[Si](C)(C)N1C(CC1=O)C(=O)O

Specifications

Melting Point 137.0°C to 141.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Formula Weight 229.35
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material (4R)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.