missing translation for 'onlineSavingsMsg'
Learn More

(4R-cis)-6-Chloromethyl-2,2-dimethyl-1,3-dioxane-4-acetic Acid tert-Butyl Ester, TRC

Product Code. 30357841
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357841 250 mg 250mg
30350637 500 mg 500mg
30350769 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357841 Supplier Toronto Research Chemicals Supplier No. TRCC369175250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 154026-94-5
Molecular Formula C13 H23 Cl O4
Molecular Weight (g/mol) 278.77
Synonym Pitavastatin Impurity 20
IUPAC Name tert-butyl 2-[(4R,6S)-6-(chloromethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
SMILES CC(C)(C)OC(=O)C[C@H]1C[C@@H](CCl)OC(C)(C)O1

Especificaciones

Quantity 250 mg
InChI Formula InChI=1S/C13H23ClO4/c1-12(2,3)18-11(15)7-9-6-10(8-14)17-13(4,5)16-9/h9-10H,6-8H2,1-5H3/t9-,10+/m1/s1
Formula Weight 278.1285
Chemical Name or Material (4R-cis)-6-Chloromethyl-2,2-dimethyl-1,3-dioxane-4-acetic acid tert-butyl ester
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.