missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4R,6R)-6-[2-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]ethyl]-4-hydroxytetrahydro-2H-pyran-2-one (Atorvastatin Lactone), Mikromol™
Description
(4R,6R)-6-[2-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]ethyl]-4-hydroxytetrahydro-2H-pyran-2-one (Atorvastatin Lactone)
Specifications
Specifications
| Chemical Name or Material | (4R,6R)-6-[2-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]ethyl]-4-hydroxytetrahydro-2H-pyran-2-one (Atorvastatin Lactone) |
| CAS | 125995-03-1 |
| Analyte or Component Names | (4R,6R)-6-[2-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]ethyl]-4-hydroxytetrahydro-2H-pyran-2-one (Atorvastatin Lactone) |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C33 H33 F N2 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Atorvastatin Calcium Trihydrate Imp. H (EP), Atorvastatin Imp. H (EP), Atorvastatin USP Related Compound H, Atorvastatin USP RC H, (4R,6R)-6-[2-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]ethyl]-4-hydroxytetrahydro-2H-pyran-2-one, Atorvastatin Lactone, Atorvastatin Related Compound H, Atorvastatin Calcium Trihydrate Impurity H, Atorvastatin Impurity H |
| InChI Formula | InChI=1S/C33H33FN2O4/c1-21(2)31-30(33(39)35-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)32(23-13-15-24(34)16-14-23)36(31)18-17-27-19-26(37)20-28(38)40-27/h3-16,21,26-27,37H,17-20H2,1-2H3,(H,35,39)/t26-,27-/m1/s1 |
| SMILES | CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H]5C[C@@H](O)CC(=O)O5 |
| IUPAC Name | 5-(4-fluorophenyl)-1-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-N,4-diphenyl-2-propan-2-ylpyrrole-3-carboxamide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?