missing translation for 'onlineSavingsMsg'
Learn More

(4R)-4,6-Dihydroxy-2-methyl-1,2,3,4-tetrahydroisoquinoline-1-carboxylic Acid (Impurity) (Mixture of 2 Diastereomers), TRC

Product Code. 30384308
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384308 100 mg
30388271 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384308 Supplier Toronto Research Chemicals Supplier No. TRCD453205100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C11 H13 N O4
Molecular Weight (g/mol) 223.225
IUPAC Name (4R)-4,6-dihydroxy-2-methyl-3,4-dihydro-1H-isoquinoline-1-carboxylic acid
SMILES CN1C[C@H](O)c2cc(O)ccc2C1C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H13NO4/c1-12-5-9(14)8-4-6(13)2-3-7(8)10(12)11(15)16/h2-4,9-10,13-14H,5H2,1H3,(H,15,16)/t9-,10?/m0/s1
Formula Weight 223.084
Chemical Name or Material (4R)-4,6-Dihydroxy-2-methyl-1,2,3,4-tetrahydroisoquinoline-1-carboxylic Acid (Impurity) (Mixture of 2 Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.