missing translation for 'onlineSavingsMsg'
Learn More

(4R)-3-[(2R,5S)-5-(2-Fluorophenyl)-2-[(S)-[(4-fluorophenyl)amino][4-[(trimethylsilyl)oxy]phenyl]methyl]-1-oxo-5-[(trimethylsilyl)oxy]pentyl]-4-phenyl-2-oxazolidinone, TRC

Product Code. 30389525
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389525 25 mg 25mg
30391560 10 mg 10mg
30394295 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389525 Supplier Toronto Research Chemicals Supplier No. TRCF59845025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4R)-3-[(2R,5S)-5-(2-Fluorophenyl)-2-[(S)-[(4-fluorophenyl)amino][4-[(trimethylsilyl)oxy]phenyl]methyl]-1-oxo-5-[(trimethylsilyl)oxy]pentyl]-4-phenyl-2-oxazolidinone
Analyte or Component Names (4R)-3-[(2R,5S)-5-(2-Fluorophenyl)-2-[(S)-[(4-fluorophenyl)amino][4-[(trimethylsilyl)oxy]phenyl]methyl]-1-oxo-5-[(trimethylsilyl)oxy]pentyl]-4-phenyl-2-oxazolidinone
Shipping Condition Room Temperature
Molecular Formula C39 H46 F2 N2 O5 Si2
InChI Formula InChI=1S/C39H46F2N2O5Si2/c1-49(2,3)47-31-22-16-28(17-23-31)37(42-30-20-18-29(40)19-21-30)33(24-25-36(48-50(4,5)6)32-14-10-11-15-34(32)41)38(44)43-35(26-46-39(43)45)27-12-8-7-9-13-27/h7-23,33,35-37,42H,24-26H2,1-6H3/t33-,35-,36+,37-/m1/s1
Solution Type Neat
SMILES C[Si](C)(C)O[C@@H](CC[C@H]([C@H](Nc1ccc(F)cc1)c2ccc(O[Si](C)(C)C)cc2)C(=O)N3[C@H](COC3=O)c4ccccc4)c5ccccc5F
IUPAC Name (4S)-3-[(2R,5S)-2-[(S)-(4-fluoroanilino)-(4-trimethylsilyloxyphenyl)methyl]-5-(2-fluorophenyl)-5-trimethylsilyloxypentanoyl]-4-phenyl-1,3-oxazolidin-2-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 716.961
Quantity 25 mg
Formula Weight 716.291
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.