missing translation for 'onlineSavingsMsg'
Learn More

(4R, 2S)-4-Hydroxy-1-(9-phenyl-9H-fluoren-9-yl)-proline Methyl Ester, TRC

Product Code. 30394358
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394358 250 mg 250mg
30404948 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394358 Supplier Toronto Research Chemicals Supplier No. TRCH949230250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179990-59-1
Molecular Formula C25 H23 N O3
Molecular Weight (g/mol) 385.46
IUPAC Name methyl (2S,4R)-4-hydroxy-1-(9-phenylfluoren-9-yl)pyrrolidine-2-carboxylate
SMILES COC(=O)[C@@H]1C[C@@H](O)CN1C2(c3ccccc3)c4ccccc4c5ccccc25

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C25H23NO3/c1-29-24(28)23-15-18(27)16-26(23)25(17-9-3-2-4-10-17)21-13-7-5-11-19(21)20-12-6-8-14-22(20)25/h2-14,18,23,27H,15-16H2,1H3/t18-,23+/m1/s1
Formula Weight 385.1678
Chemical Name or Material (4R, 2S)-4-Hydroxy-1-(9-phenyl-9H-fluoren-9-yl)-proline Meth
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.