Learn More
Jenway™ 4M KCl/AgCl Gel Fill Solution
Product Code. 11419469
Click to view available options
Type:
Partial Gel
Unit Size:
Each
Product Code. 11419469
Brand: Jenway™ 9023860
Ensures accurate and reliable analytical measurements. Jenway™ 4M KCL/AgCl Gel Fill Solution promotes optimal electrode performance when used as part of a regular care and maintenance regime.
  • For use with care and maintenance of pH electrodes
  • 4M KCl saturated with AgCl gel
  • 150mL
Specifications
Type Partial Gel
Filling Solution Type 4M KCl/AgCl
Quantity 150 mL