missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4H-Furo[3,2-c]pyranyl Mupirocin Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 4H-Furo[3,2-c]pyranyl Mupirocin Methyl Ester |
| Molecular Formula | C27H46O9 |
| Quantity | 1 mg |
| Synonym | [2R-[2α(1S*,2S*,3S*),3aβ,6α(E),7β,7aα]]-9-[[4-[2-(1,3-Dihydroxy-2-methylbutyl)hexahydro-7-hydroxy-4H-furo[3,2-c]pyran-6-yl]-3-methyl-1-oxo-2-butenyl]oxy]nonanoic Acid Methyl Ester, |
| InChI Formula | InChI=1S/C8H15NO2/c1-3-6(2)7-4-11-5-8(10)9-7/h6-7H,3-5H2,1-2H3,(H,9,10)/t6?,7-/m1/s1 |
| SMILES | CCC(C)[C@@H]1NC(=O)COC1 |
| Molecular Weight (g/mol) | 514.65 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?