missing translation for 'onlineSavingsMsg'
Learn More

4F 4PP oxalate, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18094912
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18094912 10 mg 10mg
18034052 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18094912 Supplier Tocris Bioscience Supplier No. 0523/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective 5-HT2A antagonist

4F 4PP oxalate is a selective 5-HT2A antagonist with almost as high affinity (Ki = 5.3 nM) as Ketanserin (Cat. No. 0908) but with a much lower affinity for 5-HT2C sites (Ki = 620 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144734-36-1
Molecular Formula C24H28FNO5
Molecular Weight (g/mol) 429.488
InChI Key VUJYJCRJPFMHEM-UHFFFAOYSA-N
Synonym 4f 4pp oxalate, 4-4-fluorobenzoyl-1-4-phenylbutyl piperidine oxalate, 4-fluorophenyl 1-4-phenylbutyl piperidin-4-yl methanone ethanedioate
PubChem CID 24745966
IUPAC Name (4-fluorophenyl)-[1-(4-phenylbutyl)piperidin-4-yl]methanone;oxalic acid
SMILES C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCCCC3=CC=CC=C3.C(=O)(C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 429.49
Percent Purity >99%
Chemical Name or Material 4F 4PP oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.