missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 4EGI-1

Product Code. 30002065
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002065 50 mg 50mg
30002056 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002065 Supplier Tocris Bioscience™ Supplier No. 4800/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits eIF4E:eIF4G subunit interaction

Inhibitor of eIF4E:eIF4G interaction; prevents eIF4E binding to eIF4G. Binds eIF4E and inhibits cap-dependent translation but not initiation factor-independent translation. Exhibits activity against Jurkat and A549 cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 315706-13-9
Molecular Formula C18H12Cl2N4O4S

Specifications

Quantity 50 mg
Formula Weight 451.28
Purity >98%
Molecular Weight (g/mol) 451.28
Chemical Name or Material Protein-synthesizing GTPase Compounds
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.