missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4EBP1 Polyclonal Antibody
Description
4EBP1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | 4EBP1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | EIF4EBP1 |
| Gene Accession No. | Q13541, Q60876 |
| Gene Alias | 4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1 |
| Gene Symbols | EIF4EBP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?