missing translation for 'onlineSavingsMsg'
Learn More

4E2RCat, MedChemExpress

Product Code. 16483138
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483138 10 mg 10mg
16460689 1 mg 1mg
16429696 50 mg 50mg
16425877 5 mg 5mg
16414436 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16483138 Supplier Medchem Express Supplier No. HY100733
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 4E2RCat is an inhibitor of eIF4E-eIF4G interaction with an IC50 of 13.5 μM.

4E2RCat is an inhibitor of eIF4E-eIF4G interaction with an IC50 of 13.5 μM.

Specifications

Chemical Name or Material 4E2RCat
CAS 432499-63-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H14ClNO4S2
Quantity 10 mg
Solubility Information DMSO : 23.33 mg/mL (51.17 mM; Need ultrasonic)
SMILES O=C(O)C1=CC(C2=CC=C(/C=C(SC(N3CC4=CC=CC=C4)=S)/C3=O)O2)=CC=C1Cl
Molecular Weight (g/mol) 455.93
Formula Weight 455.93
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.