missing translation for 'onlineSavingsMsg'
Learn More

4E1RCat, MedChemExpress

Product Code. 16478667
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478667 5 mg 5mg
16416068 10 mg 10mg
16402207 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478667 Supplier Medchem Express Supplier No. HY14427
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 4E1RCat is an inhibitor of cap-dependent translation, and inhibits eIF4E:eIF4GI interaction, with an IC50 an of ∼4 μM.

4E1RCat is an inhibitor of cap-dependent translation, and inhibits eIF4E:eIF4GI interaction, with an IC50 an of ∼4 μM.

Specifications

Chemical Name or Material 4E1RCat
CAS 328998-25-0
Color Dark Brown
Purity Grade Notes Research
Molecular Formula C28H18N2O6
Quantity 5 mg
Solubility Information DMSO : 18.33 mg/mL (38.31 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(N2C(/C(C=C2C3=CC=CC=C3)=C\C4=CC=C(C5=CC=C([N+]([O-])=O)C=C5)O4)=O)C=C1
Molecular Weight (g/mol) 478.45
Formula Weight 478.45
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.