missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4E-BP3 Monoclonal Antibody (1E3)

4E-BP3 Monoclonal Antibody (1E3)
Brand: Invitrogen MA520460
This item is not returnable.
View return policy
Description
4E-BP3 Monoclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
| 4E-BP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| O60516 | |
| EIF4EBP3 | |
| EIF4EBP3 (AAH10881, 1 a.a. ~ 100 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 1E3 | |
| Unconjugated | |
| EIF4EBP3 | |
| 4EBP3; 4E-BP3; eIF4E-binding protein 3; EIF4EBP3; eukaryotic initiation factor 4E-binding protein 3; eukaryotic translation initiation factor 4E binding protein 3; eukaryotic translation initiation factor 4E-binding protein 3; NG36 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8637 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction