missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 4E-BP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589361
This item is not returnable.
View return policy
Description
Immunogen sequence: DRRNSPMAQT PPCHLPNIPG VTSPGTLIED SKVEVNNLNN LNNHDRKHAV GDDAQFEMDI; Positive Samples: HeLa, Jurkat.
PHAS-II (eIF4E-binding protein II) is involved in translation initiation. PHAS-II is highest in liver and kidney. PHAS proteins bind to eIF-4E, the mRNA cap-binding protein, and inhibit translation of capped mRNA in vitro and in cells. Phosphorylation in the appropriate site(s) promotes dissociation of PHAS/eIF-4E complexes. This allows eIF-4E to bind to eIF-4G (p220), thereby increasing the amount of the eIF-4F complex and the rate of translation initiation. Pharmacological and genetic evidence indicates that the mTOR/p70S6K pathway is involved in the control of PHAS-I and -II. Thus, PHAS proteins may be mediators of the effects of this pathway on protein synthesis and cell proliferation.
Specifications
| 4E-BP2 | |
| Polyclonal | |
| Unconjugated | |
| EIF4EBP2 | |
| 2810011I19Rik; 4EBP2; 4E-BP2; AA792569; BC010348; eIF4E-binding protein 2; EIF4EBP2; eukaryotic translation initiation factor 4E binding protein 2; eukaryotic translation initiation factor 4E-binding protein 2; PHASII; PHAS-II; Phosphorylated heat- and acid-stable protein regulated by insulin 2; phosphorylated, heat and acid stable regulated by insulin protein II | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1979 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.45 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q13542 | |
| EIF4EBP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 61-120 of human EIF4EBP2 (NP_0040871). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction