missing translation for 'onlineSavingsMsg'
Learn More

4Beta-Hydroxy Stanozolol, TRC

Product Code. 30399251
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399251 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399251 Supplier Toronto Research Chemicals Supplier No. TRCH95355510MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4b-Hydroxystanozolol
CAS 125636-92-2
Molecular Formula C21 H32 N2 O2
Quantity 10 mg
Synonym 2'H-Androst-2-eno[3,2-c]pyrazole-4,17-diol, 17-methyl-, (4β,5α,17β)- (9CI, ACI), (4β,5α,17β)-17-Methyl-2'H-androst-2-eno[3,2-c]pyrazole-4,17-diol (ACI), Cyclopenta[7,8]phenanthro[2,3-c]pyrazole, 2'H-androst-2-eno[3,2-c]pyrazole-4,17-diol deriv. (ZCI), 4β-Hydroxystanozolol
InChI Formula InChI=1S/C21H32N2O2/c1-19-10-12-11-22-23-17(12)18(24)16(19)5-4-13-14(19)6-8-20(2)15(13)7-9-21(20,3)25/h11,13-16,18,24-25H,4-10H2,1-3H3,(H,22,23)/t13-,14+,15+,16+,18-,19-,20+,21+/m1/s1
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4[C@@H](O)c5[nH]ncc5C[C@]4(C)[C@H]3CC[C@]12C
Molecular Weight (g/mol) 344.49
Formula Weight 344.2464
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.