missing translation for 'onlineSavingsMsg'
Learn More

(4aS,6R,7S,7aR)-6-[4-[[(1S)-2,3-Dihydro-1H-inden-1-yl]amino]-7H-pyrrolo[2,3-d]pyrimidin-7-yl]hexahydro-2-(4-methoxyphenyl)-cyclopenta-1,3-dioxin-7-ol, TRC

Product Code. 30388436
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388436 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388436 Supplier Toronto Research Chemicals Supplier No. TRCD44989010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4aS,6R,7S,7aR)-6-[4-[((1S)-2,3-Dihydro-1H-inden-1-yl)amino]-7H-pyrrolo[2,3-d]pyrimidin-7-yl]hexahydro-2-(4-methoxyphenyl)-cyclopenta-1,3-dioxin-7-ol
CAS 905580-87-2
Molecular Formula C29H30N4O4
Quantity 10 mg
Synonym (4As,6R,7S,7Ar)-6-(4-((S)-2,3-Dihydro-1H-Inden-1-Ylamino)-7H-Pyrrolo[2,3-D]Pyrimidin-7-Yl)-2-(4-Methoxyphenyl)Hexahydrocyclopenta[D][1,3]Dioxin-7-Ol
InChI Formula InChI=1S/C29H30N4O4/c1-35-20-9-6-18(7-10-20)29-36-15-19-14-24(25(34)26(19)37-29)33-13-12-22-27(30-16-31-28(22)33)32-23-11-8-17-4-2-3-5-21(17)23/h2-7,9-10,12-13,16,19,23-26,29,34H,8,11,14-15H2,1H3,(H,30,31,32)/t19-,23-,24+,25-,26+,29?/m0/s1
SMILES COc1ccc(cc1)C2OC[C@@H]3C[C@H]([C@H](O)[C@@H]3O2)n4ccc5c(N[C@H]6CCc7ccccc67)ncnc45
Molecular Weight (g/mol) 498.57
Product Title
Select an issue

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.