missing translation for 'onlineSavingsMsg'
Learn More

4alpha-Phorbol 12-myristate 13-acetate, 99%, Thermo Scientific™

Product Code. 15483749
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 15483749

Brand: Thermo Scientific Alfa Aesar J61099.M^

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Negative control for phorbol ester activation of protein kinase C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4alpha-Phorbol 12-myristate 13-acetate is a non-tumor promoting phorbol ester derivative that has been used as a negative control for PMA, a tumor promoter and activator of PKC. However, 4α-PMA has also been shown to be a reasonably potent activator of TRPV4 channels, making it a useful tool for structure-activity investigations via comparison to the very similar compound, 4α-PDD, which demonstrates 50 times more affinity for TRPV4. Furthermore, this compound has also been shown to increase PGHS-2 mRNA significantly in murine keratinocytes.

Solubility
Soluble in DMSO, methanol or ethanol. Dilute stock solution into aqueous medium at final concentrations of ∼2-5µM

Notes
Light sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63597-44-4
Molecular Formula C36H56O8
Molecular Weight (g/mol) 616.84
MDL Number MFCD00153860
InChI Key PHEDXBVPIONUQT-LQLWEASQSA-N
Synonym 4alpha-phorbol 12-myristate 13-acetate, phorbol-12-myristate-13-acetate, pma, 4alpha-pma, 4alpha-tpa, .alpha.-tpa, 4.alpha.-tpa, 4alpha-phorbol-12-myristate-13-acetate, 4, a-pma, acetoxy-dihydroxy-hydroxymethyl-tetramethyl-oxo-? yl tetradecanoate
PubChem CID 122634
IUPAC Name (1S,2S,6S,10S,11R,13S,14R,15R)-13-(acetyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-14-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(C)=O

Specifications

Color White
Quantity 1 mg
Sensitivity Light sensitive
Merck Index 14,7332
Solubility Information Soluble in DMSO,methanol or ethanol; Dilute stock solution into aqueous medium at final concentrations of ∽2-5μM
Formula Weight 616.83
Percent Purity 99%
Physical Form Film or Powder
Chemical Name or Material 4alpha-Phorbol 12-myristate 13-acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.