missing translation for 'onlineSavingsMsg'
Learn More

4Alpha-Hydroxy Stanozolol, TRC

Product Code. 30399372
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30399372 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399372 Supplier Toronto Research Chemicals Supplier No. TRCH9535451MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 4a-Hydroxystanozolol
CAS 100356-20-5
Molecular Formula C21 H32 N2 O2
Quantity 1 mg
Synonym 4alpha-Hydroxy Stanozolol, (4α,5α,17β)-17-Methyl-2'H-androst-2-eno[3,2-c]pyrazole-4,17-diol, 1,2,3,3a,3b,4,5,5a,6,8,10,10a,10b,11,12,12a-Hexadecahydro-1,10a,12a-trimethylcyclopenta[7,8]phenanthro[2,3-c]pyrazole-1,6-diol, 4α-Hydroxystanazolol, 4α-Hydroxystanozolol
InChI Formula InChI=1S/C21H32N2O2/c1-19-10-12-11-22-23-17(12)18(24)16(19)5-4-13-14(19)6-8-20(2)15(13)7-9-21(20,3)25/h11,13-16,18,24-25H,4-10H2,1-3H3,(H,22,23)/t13-,14+,15+,16+,18+,19-,20+,21+/m1/s1
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4[C@H](O)c5[nH]ncc5C[C@]4(C)[C@H]3CC[C@]12C
Molecular Weight (g/mol) 344.4
Formula Weight 344.491
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.