missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 7 2000 D MWCO Standard RC Pre-Treated Dialysis Kits

Product Code. 15320692
Change view
Click to view available options
Diameter (Metric):
11.5 mm
24 mm
29 mm
Length (Metric):
1.1 mL/cm vol./length
4.6 mL/cm vol./length
6.4 mL/cm vol./length
Unit Size:
1m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Unit Size
15320692 24 mm 4.6 mL/cm vol./length 1m
15310692 11.5 mm 1.1 mL/cm vol./length 1m
15330692 29 mm 6.4 mL/cm vol./length 1m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15320692 Supplier Spectrum™ Supplier No. 132108T
  • ONLINE EXCLUSIVE PRICE
    £162.00
    £153.73 / 1m
    Save £8.27
    5% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • 1meter of pre-treated tubing, a pair of closures and 5 opening picks

With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds

  • Hydrophilic, symmetric porositiy
  • Equilibrated in water
  • Packaged wet in preservative solution of 0.05% sodium azide
  • For small experiments or qualifying a membrane for a particular application

Specifications

Product Type Dialysis Trial Kit
MWCO 2 kDa
Format Tubing
Diameter (Metric) 24 mm
Length (Metric) 4.6 mL/cm vol./length
Quantity 1 m Tubing/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.