missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Biohazard Identification Signs: Biohazard Infectious Agent Entry Requirements Contact Phone

Product Code. 16421481
Click to view available options
Material:
Aluminum
Plastic
Polyester
This item is not returnable. View return policy

Product Code. 16421481

Brand: Brady™ 126647

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For identification of equipment, rooms and materials that contain, or are contaminated with, hazardous agents

  • Made from aluminum, plastic or self-sticking polyester
  • Size: 14 in. H x 10 in. W
  • Legend: BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___

Specifications

Material Polyester
Description Self-sticking Polyester
Height (English) 14 in.
Height (Metric) 4.2 m
Width (English) 10 in.
Width (Metric) 3 m
Displayed Text BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___
Product Type Biohazard Identification Sign
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.