missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Biohazard Identification Signs: Biohazard Infectious Agent Entry Requirements Contact Phone
Description
- Made from aluminum, plastic or self-sticking polyester
- Size: 14 in. H x 10 in. W
- Legend: BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___
Specifications
Specifications
| Material | Plastic |
| Height (English) | 14 in. |
| Height (Metric) | 4.2 m |
| Width (English) | 10 in. |
| Width (Metric) | 3 m |
| Displayed Text | BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___ |
| Product Type | Biohazard Identification Sign |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?