missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Biohazard Identification Signs: Biohazard Infectious Agent Entry Requirements Contact Phone
Click to view available options
Material:
Aluminum
Plastic
Polyester
Description
- Made from aluminum, plastic or self-sticking polyester
- Size: 14 in. H x 10 in. W
- Legend: BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___
Specifications
Specifications
| Material | Aluminum |
| Height (English) | 14 in. |
| Height (Metric) | 4.2 m |
| Width (English) | 10 in. |
| Width (Metric) | 3 m |
| Displayed Text | BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___ |
| Product Type | Biohazard Identification Sign |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction