Learn More
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
Brand:  Hamilton™ 7640-01
Code : A0
Additional Details : Weight : 0.03300kg
 View more versions of this product
Product Code. 11577864
  • £143.00 / Each
Estimated Shipment: 16-04-2024
to see stock.
Description
Description
  • ideal for dispensing volumes from 0.05 µL up to 5 µL
  • The 7000 series is the only zero dead volume syringe
  • The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle
Specifications
Specifications
22 G
750
500 μL