missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates™ immunoGrade™ Strip Plates
Immunological surfaces for immobilization of IgG
£471.00 - £660.00
Specifications
Format | Strip Plate |
---|---|
Array | 1 x 8 |
Binding Type | High Binding |
Color | Clear |
No. of Wells | 8 |
Product Code | Brand | Total Volume (Metric) Well | Breakapart | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Total Volume (Metric) Well | Breakapart | Price | Quantity & Availability | ||||
11663189
|
BRAND™
782306 |
350 μL | Yes |
£660.00
Pack of 100 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
11653189
|
BRAND™
782305 |
360 μL | No |
£471.00
Pack of 100 |
Estimated Shipment: 27-06-2024
Log in to see stock. |
||||
Description
- High binding and non-sterile
- Made from polystyrene
- Transparent with flat bottom
- With and without grid
- Free from endotoxins, DNase, DNA, RNase; non-cytotoxic
- Modification via physicalchemical process
With grid: Individual strips can be removed from the plate and reinserted, the strips can be divided into 8 individual vessels and inserted individually into the frame plate.
Without grid: individual strips can be removed from the frame and reinserted, the strips are not divisible.
Applications:
For solid phase immunoassays, ELISA, RIA, FIA and other immune testing
Specifications
Strip Plate | |
High Binding | |
8 | |
Immunological | |
100/Pk. |
1 x 8 | |
Clear | |
Non-sterile | |
Flat Bottom (F-well) |