Learn More
BioTek™ 48-tube Rack for Test Tubes
48-tube Rack for 12 to 13 mm Test Tubes is for use with BioTek™ Microplate Washers and Readers
Brand: BioTek™ 7110538
Additional Details : Weight : 0.50000kg
Specifications
Tube Rack | |
48 Tubes | |
BioTek™ Microplate Washer and Reader Instruments | |
12 to 13 mm |