Learn More
3M™ 4432+ Lab Coats
Product Code. p-8263042
Change view
Click to view available options
Size:
X-Large
2X-Large
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15388766 2X-Large Pack of 50
15388756 X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15388766 Supplier 3M™ Supplier No. 7100040414
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to help protect the wearer against light splashing with non-hazardous liquids and dust. 3M™ 4432+ Lab Coats are made from super-breathable SSMMS non-woven for effective release of body heat and moisture.
  • Anti-static
  • Front zip closing
  • Low lint properties
  • Reduced heat stress
  • Knitted cuffs and collared
  • Internal pocket on the wearer's left-hand side
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material Polypropylene, Polyethylene Laminate
Color Cool White
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Seam Type Serged
Certifications/Compliance CE Category 1
Packaging Type Single Pack