missing translation for 'onlineSavingsMsg'
Learn More

42-O-[2-[[Hydroxy[2-(trimethylammonio)ethoxy]phosphinyl]oxy]ethyl] Rapamycin Inner Salt >75%, TRC

Product Code. 30392542
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392542 1 mg
30400132 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392542 Supplier Toronto Research Chemicals Supplier No. TRCH9721001MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1062645-51-5
Molecular Formula C58H95N2O17P
Molecular Weight (g/mol) 1123.35
Synonym ATG 181
SMILES C[C@@H]1CC[C@H]2C[C@H](/C(=C/C=C/C=C/[C@H](C[C@H](C(=O)[C@@H]([C@@H](/C(=C/[C@H](C(=O)C[C@H](OC(=O)[C@@H]3CCCCN3C(=O)C(=O)[C@@]1(O2)O)[C@H](C)C[C@@H]4CC[C@H]([C@@H](C4)OC)OCCOP(=O)([O-])OCC[N+](C)(C)C)C)/C)O)OC)C)C)/C)OC

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C58H95N2O17P/c1-37-19-15-14-16-20-38(2)49(70-11)35-45-24-22-43(7)58(67,77-45)55(64)56(65)59-26-18-17-21-46(59)57(66)76-50(36-47(61)39(3)32-42(6)53(63)54(72-13)52(62)41(5)31-37)40(4)33-44-23-25-48(51(34-44)71-12)73-29-30-75-78(68,69)74-28-27-60(8,9)10/h14-16,19-20,32,37,39-41,43-46,48-51,53-54,63,67H,17-18,21-31,33-36H2,1-13H3/b16-14+,19-15+,38-20+,42-32+/t37-,39-,40-,41-,43-,44+,45+,46+,48-,49-,50+,51-,53-,54+,58-/m1/s1
Formula Weight 1122.6368
Chemical Name or Material 42-O-[2-[(Hydroxy[2-(trimethylammonio)ethoxy]phosphinyl)oxy]ethyl] Rapamycin Inner Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.