missing translation for 'onlineSavingsMsg'
Learn More
Learn More
40kDa Pan Carcinoma Mouse anti-Human, BB700, Clone: KS1/4, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 745829
This item is not returnable.
View return policy
Description
40kDa Pan Carcinoma Monoclonal antibody specifically detects 40kDa Pan Carcinoma in Human samples. It is validated for Flow Cytometry.
Specifications
| 40kDa Pan Carcinoma | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| KS1/4 | |
| Brilliant Blue 700 (BB700) | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction