missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMS™ Series 3 Model 400WNP Cooled Incubator, 450 L, Stainless Steel
Description
- Range of mid-sized benchtop and freestanding cooled incubators with a white stove enamel mild steel exterior, foam insulation and stainless steel interiors
- Reliable and accurate temperature control
- Standard single and dual temperature, plus programmable models available
- Temperature range -10.0°C to +50.0°C, temperature variation with average workload±0.5°C
- Self-evaporating condensate drip tray
- White plastic coated wire shelves
- Energy efficient control system and eco-friendly
- P.I.D. heating and cooling to ensure stable internal temperature
- Pre-set safety cutout
- Hermetically sealed refrigeration system
- Door lock
- Digital controls are easy to read and simple to use
- Full range offactory-fitted optional extras available, including LED lighting, access porthole, alarm system, inner perspex door, internal socket and chart recorder
- Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2008 |
| Controller Type | Programmable |
| Description | Freestanding incubator |
| Capacity (Metric) | 450 L |
| Chamber Material | Stainless Steel |
| Convection Type | Fan-assisted Circulation |
| Dimensions (D x W) Shelf | 510 mm x 600 mm |
| Lighting | Optional Fluorescent/UV |
| Material | Stove Enameled Mild Steel |
| No. of Shelves | 3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
