missing translation for 'onlineSavingsMsg'
Learn More

Acid Number Standard 0.1 mgKOH/g. Reference materials intended for use in the determination of Acid, VHG™

Product Code. 17963926
Click to view available options
Quantity:
100 g
400 g
800 g
Unit Size:
100g
400g
800g
This item is not returnable. View return policy

Product Code. 17963926

Brand: VHG Labs VHGAN0.1400G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Acid Number Standard 0.1 mgKOH/g. Reference materials intended for use in the determination of Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium hydroxide
CAS 1310-58-3
Methods Potentiometric Titration
Analyte or Component Names Potassium hydroxide
Shipping Condition Room Temperature
Molecular Formula H K O
Concentration 0.1 mg/g
Certifications/Compliance ISO 17034
Synonym Potassium hydroxide
InChI Formula InChI=1S/K.H2O/h;1H2/q+1;/p-1
SMILES O[K]
IUPAC Name hydroxypotassium
Solvent or Matrix Hydrocarbon Oil
Molecular Weight (g/mol) 56.11
Quantity 400 g
Formula Weight 55.9664 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.