missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ 4000 Series Disposable Respirator, FFP1 (NR D)

Product Code. 11891771
Change view
Click to view available options
Honeywell™ Disposable Respirator; Filter Efficiency: FFP1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11891771 Honeywell™ Disposable Respirator; Filter Efficiency: FFP1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11891771 Supplier Honeywell™ Supplier No. 1005605
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The 4000 Series Disposable Respirator is a single-use, filtering, flat-fold, half-mask with a fluid-resistant outer shell and elastic fasteners. With no exhalation valve it has a Willtech™ nasal seal. Supplied in an individual bag for maintenance of hygiene.

  • Product Type: General Purpose Particulate Respirator
  • Description: Half-Mask
  • Shape: Flat-Fold
  • Model: 4110
  • Filter Efficiency: FFP1
  • Size: M/L
  • Seal Type: Nasal Seal (Willtech™)
  • Disposable: Yes
  • Latex Free: Yes
  • Resistant To: Fluid
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EC Category: 3, EN 149:2001+A1:2009
Color White
Design Type Flat-fold
Disposable Single-use
Material Polyester
Product Type General Purpose Particulate Respirator
Size Medium/Large
With Exhalation Valve No
Filter Efficiency FFP1
For Use With (Application) Textile, Construction, Iron and Steel Industries
Latex Free Yes
Material (Faceshield) PES Knit, PU Foam, PU Film
Material Headstrap Elastic
Model 4110
Nose Piece Material Polypropylene-wrapped Galvanized Wire
Packaging Type Individual Bagged
Quantity 20 Pk.
Resistant To Fluid
Sterility Non-sterile
Style Half-Mask, Single-Use
With Faceseal Yes (Nasal Seal, Willtech™)
Weight (Metric) 7 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.