Learn More
TubeWriter 400 - Standard Rack Side Print for 50 mL Conical Centrifuge Tubes
Product Code. 30067440
Change view
Click to view available options
Product Type:
Labeling Rack
Unit Size:
Each
Product Code. Product Type Unit Size
30067440 Labeling Rack Each
1 options
Product Code. 30067440 Supplier TubeWriter Supplier No. PS0050CNEL120400
  • ONLINE EXCLUSIVE PRICE
    £759.00
    £741.84 / Pack of 1
    Save £17.16
    2% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Standard rack for directly printing on the side of 50 mL conical centrifuge tubes
  • Standard printing rack designed for the 400
  • Prints and labels directly on the side of 50 mL conical centrifuge tubes
  • Rack ensures correct alignment and holds tubes securely
  • No time-consuming sticky labels or handwriting required
  • Printing can take as little time as 50 tubes in 2 to 3 minutes
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80C and liquid nitrogen
  • Print withstands the harshest lab conditions
Specifications
Product Type Labeling Rack