missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RETSCH 400 dia. x 65mmH Stainless Steel Test Sieve, ISO Certified
Brand: RETSCH 60.166.000560
Additional Details : Weight : 1.60000kg
Specifications
For separation, fractioning, particle size determination | |
Stainless Steel | |
1Each | |
Test Sieve | |
Stainless Steel | |
65mm |
400mm | |
Stainless Steel | |
560μm | |
ISO 3310/1 | |
Shaker AS 450 |