missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylpyridine, 95%, stab. with 80 to 150ppm hydroquinone

Product Code. p-7046152
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 11388026

Brand: Thermo Scientific Alfa Aesar L13316.AC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-43-6 | C7H7N | 105.14 g/mol

4-Vinylpyridine acts as a co-monomer in styrene-butadiene polymers to promote adhesion between the rubber compound and supporting fibers (or cords) in tires, belts and hoses. It serves as an intermediate for manufacturing chemicals and pharmaceuticals. Further, it is used as a reagent for the modification of cysteine. In addition to this, it is used to make poly(4-vinylpyridine) and elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylpyridine acts as a co-monomer in styrene-butadiene polymers to promote adhesion between the rubber compound and supporting fibers (or cords) in tires, belts and hoses. It serves as an intermediate for manufacturing chemicals and pharmaceuticals. Further, it is used as a reagent for the modification of cysteine. In addition to this, it is used to make poly(4-vinylpyridine) and elastomers.

Solubility
Miscible with water, chloroform, ethanol, dilute acids, alcohol, hydrocarbons, esters and ketones.

Notes
Light sensitive. Incompatible with strong acids, strong oxidizing agents, ultraviolet radiation and free radical initiators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-43-6
Molecular Formula C7H7N
Molecular Weight (g/mol) 105.14
MDL Number MFCD00006447
InChI Key KFDVPJUYSDEJTH-UHFFFAOYSA-N
Synonym 4-vinylpyridine, pyridine, 4-ethenyl, pyridine, 4-vinyl, gamma-vinylpyridine, unii-i56g67xm8d, ccris 5240, 4vp, 4-pyridylethylene, 4-vinyl-pyridine, 4-vinyl pyridine
PubChem CID 7502
IUPAC Name 4-ethenylpyridine
SMILES C=CC1=CC=NC=C1

Specifications

Density 0.984
Boiling Point 58°C to 61°C (12 mmHg)
Flash Point 51°C (124°F)
Refractive Index 1.551
Quantity 25 mL
UN Number UN3073
Beilstein 104506
Solubility Information Miscible with water,chloroform,ethanol,dilute acids,alcohol,hydrocarbons,esters and ketones.
Formula Weight 105.14
Percent Purity 96%
Chemical Name or Material 4-Vinylpyridine, stabilized with 100ppm hydroquinone
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Vinylpyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-Vinylpyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.