missing translation for 'onlineSavingsMsg'
Learn More

(4-Vinylphenyl)methanol (~0.1% TBC stabilizer), TRC

Product Code. 30457265
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457265 50 mg 50mg
30452195 500 mg 500mg
30454062 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457265 Supplier Toronto Research Chemicals Supplier No. TRCV45331550MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-61-9
Molecular Formula C9 H10 O
Molecular Weight (g/mol) 134.18
Synonym Benzyl alcohol, p-vinyl- (6CI,7CI,8CI), 4-Ethenylbenzenemethanol, (4-Ethenylphenyl)methanol, 4-(Hydroxymethyl)styrene, 4-Vinylbenzenemethanol, 4-Vinylbenzyl alcohol, p-(Hydroxymethyl)styrene, p-Vinylbenzyl alcohol
IUPAC Name (4-vinylphenyl)methanol
SMILES OCc1ccc(C=C)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H10O/c1-2-8-3-5-9(7-10)6-4-8/h2-6,10H,1,7H2
Formula Weight 134.0732
Chemical Name or Material 4-Vinylbenzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.