missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylphenol, min 10% soln. in propylene glycol

Product Code. 11343066
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343066 5 g 5g
11333066 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343066 Supplier Thermo Scientific Alfa Aesar Supplier No. L10902.06
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2628-17-3 | C8H8O | 120.151 g/mol

4-Vinylphenol acts as a monomer and used in the preparation of poly-4-vinylphenol polymer, which is used in electronics as a dielectric layer in organic transistors in organic thin-film-transistor liquid-crystal displays (TFT LCD). It is a phenolic compound and presents in wine and beer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylphenol acts as a monomer and used in the preparation of poly-4-vinylphenol polymer, which is used in electronics as a dielectric layer in organic transistors in organic thin-film-transistor liquid-crystal displays (TFT LCD). It is a phenolic compound and presents in wine and beer.

Solubility
Slightly miscible with water.

Notes
Hygroscopic. Moisture and heat sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2628-17-3
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00017593
InChI Key FUGYGGDSWSUORM-UHFFFAOYSA-N
Synonym 4-vinylphenol, 4-hydroxystyrene, p-vinylphenol, p-hydroxystyrene, phenol, 4-ethenyl, 4-vinylphenol, 10 wt.% in propylene glycol, phenol, p-vinyl, unii-oa7v1sm8yl, fema no. 3739, poly 4-hydroxystyrene
PubChem CID 62453
ChEBI CHEBI:1883
IUPAC Name 4-ethenylphenol
SMILES C=CC1=CC=C(C=C1)O

Specifications

Density 1.04
Flash Point >100°C (212°F)
Refractive Index 1.447
Quantity 5 g
Beilstein 506844
Sensitivity Hygroscopic
Solubility Information Slightly miscible with water.
Formula Weight 120.15
Concentration or Composition (by Analyte or Components) ≥10% soln. in propylene glycol
Chemical Name or Material 4-Vinylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.