missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbiphenyl, 97%

Product Code. 15498939
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15498939

Brand: Thermo Scientific Alfa Aesar L11869.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2350-89-2 | C14H12 | 180.25 g/mol

4-Vinylbiphenyl intermediate is used in organic synthesis and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Vinylbiphenyl intermediate is used in organic synthesis and in chemical research.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilat
TRUSTED_SUSTAINABILITY

Specifications

CAS 2350-89-2
Melting Point 115°C to 116°C
Molecular Formula C14H12
MDL Number MFCD00008620
Quantity 1 g
Beilstein 2039221
Synonym 4-vinylbiphenyl, p-vinylbiphenyl, 4-phenylstyrene, biphenyl, 4-vinyl, 1,1'-biphenyl, 4-ethenyl, p-phenylstyrene, 4-ethenyl-1,1'-biphenyl, 4-vinyldiphenyl, 4-vinyl biphenyl, 4-vinyl-biphenyl
Solubility Information Insoluble in water.
InChI Key HDBWAWNLGGMZRQ-UHFFFAOYSA-N
SMILES C=CC1=CC=C(C=C1)C1=CC=CC=C1
IUPAC Name 1-ethenyl-4-phenylbenzene
Molecular Weight (g/mol) 180.25
PubChem CID 16883
Formula Weight 180.25
Percent Purity 97%
Chemical Name or Material 4-Vinylbiphenyl
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.