missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzyl acetate, 95%, stabilized

Product Code. 10215621 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215621 5 g Glass bottle 5g
10632952 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10215621 Supplier Thermo Scientific Acros Supplier No. 422500050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1592-12-7 | C11H12O2 | 176.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1592-12-7
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD00078265
InChI Key LEIKPUSDAWATBV-UHFFFAOYSA-N
Synonym 4-vinylbenzyl acetate, 4-ethenylphenyl methyl acetate, 4-acetoxymethylstyrene, 4-vinylbenzylacetate, p-acetoxymethylstyrene, 4-vinylphenyl methyl acetate, 4-ethenylbenzenemethanol acetate, benzenemethanol,4-ethenyl-, 1-acetate
PubChem CID 2735161
IUPAC Name (4-ethenylphenyl)methyl acetate
SMILES CC(=O)OCC1=CC=C(C=C)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White
Density 1.5300g/mL
Boiling Point 125°C (12.0 mmHg)
Flash Point >93°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5350 to 1.538
Quantity 5 g
Specific Gravity 1.53
Stabilizer 0.1 % hydroquinone
Formula Weight 176.22
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Vinylbenzyl acetate, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 4-Vinylbenzyl acetate, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.