missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzyl acetate, 95%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3779890
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1592-12-7
C11H12O2
176.22
MFCD00078265
LEIKPUSDAWATBV-UHFFFAOYSA-N
4-vinylbenzyl acetate, 4-ethenylphenyl methyl acetate, 4-acetoxymethylstyrene, 4-vinylbenzylacetate, p-acetoxymethylstyrene, 4-vinylphenyl methyl acetate, 4-ethenylbenzenemethanol acetate, benzenemethanol,4-ethenyl-, 1-acetate
2735161
(4-ethenylphenyl)methyl acetate
CC(=O)OCC1=CC=C(C=C)C=C1
This item is not returnable. View return policy
1592-12-7
C11H12O2
176.22
MFCD00078265
LEIKPUSDAWATBV-UHFFFAOYSA-N
4-vinylbenzyl acetate, 4-ethenylphenyl methyl acetate, 4-acetoxymethylstyrene, 4-vinylbenzylacetate, p-acetoxymethylstyrene, 4-vinylphenyl methyl acetate, 4-ethenylbenzenemethanol acetate, benzenemethanol,4-ethenyl-, 1-acetate
2735161
(4-ethenylphenyl)methyl acetate
CC(=O)OCC1=CC=C(C=C)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color White
Density 1.5300g/mL
Boiling Point 125°C (12.0 mmHg)
Flash Point >93°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5350 to 1.538
Quantity 5 g
Specific Gravity 1.53
Stabilizer 0.1 % hydroquinone
Formula Weight 176.22
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Vinylbenzyl acetate, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 4-Vinylbenzyl acetate, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'