missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzoic acid, 98%

Product Code. 11468983
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468983 1 g 1g
11478983 5 g 5g
11488983 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468983 Supplier Thermo Scientific Alfa Aesar Supplier No. A11107.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1075-49-6 | C9H8O2 | 148.161 g/mol

It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Soluble in dichloromethane and methanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075-49-6
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00002569
InChI Key IRQWEODKXLDORP-UHFFFAOYSA-N
Synonym 4-vinylbenzoic acid, p-vinylbenzoic acid, 4-carboxystyrene, benzoic acid, 4-ethenyl, styrene-4-carboxylic acid, 4-vinyl-benzoic acid, p-vinyl benzoic acid, 4-vinyl benzoic acid, para-vinyl benzoic acid, pubchem12624
PubChem CID 14098
IUPAC Name 4-ethenylbenzoic acid
SMILES C=CC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point ∼142°C
Density 1.3
Quantity 1 g
Beilstein 2041130
Sensitivity Light sensitive
Solubility Information Soluble in dichloromethane and methanol
Formula Weight 148.16
Percent Purity 98%
Chemical Name or Material 4-Vinylbenzoic acid
compliance-icons
Product Identifier
  • 4-Vinylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: p-Vinylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.