missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzoic acid, 96%

Product Code. 10782695 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10782695 5 g Glass bottle 5g
10229653 250 mg Glass bottle 250mg
10773141 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10782695 Supplier Thermo Scientific Acros Supplier No. 294910050
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1075-49-6 | C9H8O2 | 148.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1075-49-6
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00002569
InChI Key IRQWEODKXLDORP-UHFFFAOYSA-N
Synonym 4-vinylbenzoic acid, p-vinylbenzoic acid, 4-carboxystyrene, benzoic acid, 4-ethenyl, styrene-4-carboxylic acid, 4-vinyl-benzoic acid, p-vinyl benzoic acid, 4-vinyl benzoic acid, para-vinyl benzoic acid, pubchem12624
PubChem CID 14098
IUPAC Name 4-ethenylbenzoic acid
SMILES C=CC1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 142.0°C to 144.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CH2CHC6H4COOH
Quantity 5 g
Solubility Information (10% in Methanol) Clear solution
Stabilizer 0.1% t-butylcatechol
Formula Weight 148.16
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-Vinylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Vinylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.