missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzo-18-crown-6, 97%

Product Code. 10369010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Plastic bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10369010 250 mg Plastic bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10369010 Supplier Thermo Scientific Acros Supplier No. 277472500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 39557-71-6 | C18H26O6 | 338.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 39557-71-6
CAS Min % 96.0
CAS Max % 100.0
Melting Point 57°C to 60°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Molecular Formula C18H26O6
MDL Number MFCD00143381
Quantity 250 mg
Synonym 4-vinylbenzo-18-crown-6, 18-ethenyl-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine, 18-vinyl-2,3,5,6,8,9,11,12,14,15-decahydrobenzo b 1,4,7,10,13,16 hexaoxacyclooctadecine, 4'-vinylbenzo-18-crown-6, 4 inverted exclamation marka-vinylbenzo-18-crown-6, 3,4-1,4,7,10,13,16-hexaoxahexadecane-1,16-diyl styrene, 20-ethenyl-2,5,8,11,14,17-hexaoxabicyclo 16.4.0 docosa-1 18 ,19,21-triene, 18-ethenyl-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecin, 18-vinyl-1,2,3,4,5,6,7,8,9,10,11,12,13,14,15,16-hexadecahydrobenzo a 18 annul ene
Solubility Information Solubility in water: .. Other solubilities: soluble in chloroform,benzene,methanol,acetone
InChI Key WMBFARVSOWVDII-UHFFFAOYSA-N
SMILES C=CC1=CC2=C(C=C1)OCCOCCOCCOCCOCCO2
IUPAC Name 20-ethenyl-2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(18),19,21-triene
Molecular Weight (g/mol) 338.39
PubChem CID 2724803
Formula Weight 338.39
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Vinylbenzo-18-crown-6, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.