missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylaniline, 90%, technical, stabilized

Product Code. 10484195 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10484195 25 g Glass bottle 25g
10381184 1 g Glass bottle 1g
10287080 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10484195 Supplier Thermo Scientific Acros Supplier No. 308480250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1520-21-4 | C8H9N | 119.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1520-21-4
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.17
MDL Number MFCD00015329
InChI Key LBSXSAXOLABXMF-UHFFFAOYSA-N
Synonym 4-vinylaniline, 4-aminostyrene, p-aminostyrene, benzenamine, 4-ethenyl, 4-aminostryene, ccris 4326, benzenamine,4-ethenyl, p-vinylaniline, p-amino styrene, para-aminostyrene
PubChem CID 73700
IUPAC Name 4-ethenylaniline
SMILES C=CC1=CC=C(C=C1)N

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 23°C to 24°C
Color Red to Yellow
Density 1.0100g/mL
Boiling Point 249°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC6H4NH2
Refractive Index 1.6250 to 1.629
Quantity 25 g
Specific Gravity 1.01
Formula Weight 119.17
Percent Purity 90%
Grade Technical
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Vinylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.