missing translation for 'onlineSavingsMsg'
Learn More

4-Ureidobenzeneboronic acid pinacol ester, 98%, Thermo Scientific™

Product Code. 15490326
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490326 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15490326 Supplier Thermo Scientific Alfa Aesar Supplier No. H53046.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877134-77-5
Molecular Formula C13H19BN2O3
Molecular Weight (g/mol) 262.116
MDL Number MFCD06795683
InChI Key BVWCMMVUJQHJJE-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl urea, 4-ureido phenylboronic acid, pinacol ester, 4-carbamoylamino benzeneboronic acid, pinacol ester, 4-ureidophenylboronic acid pinacol ester, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl urea, 4-n-aminocarbonyl aminophenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenylurea, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl urea, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylurea, 4-ureidobenzeneboronic acid pinacol ester
PubChem CID 16727440
IUPAC Name [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]urea
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC(=O)N

Specifications

Melting Point 124°C to 128°C
Quantity 250 mg
Formula Weight 262.12
Percent Purity 98%
Chemical Name or Material 4-Ureidobenzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.