missing translation for 'onlineSavingsMsg'
Learn More

4-[(Trimethylsilyl)ethynyl]benzaldehyde, 97%

Product Code. 15451188
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451188 25 g 25g
15431188 1 g 1g
15441188 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451188 Supplier Thermo Scientific Alfa Aesar Supplier No. H55060.14
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77123-57-0 | C12H14OSi | 202.33 g/mol

It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Notes
Store at 4°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77123-57-0
Molecular Formula C12H14OSi
Molecular Weight (g/mol) 202.33
MDL Number MFCD02093765
InChI Key UZQDUXAJFTWMDT-UHFFFAOYSA-N
Synonym 4-trimethylsilyl ethynyl benzaldehyde, 4-trimethylsilyl ethynylbenzaldehyde, 4-2-trimethylsilylethynyl benzaldehyde, 4-2-trimethylsilyl ethynyl benzaldehyde, benzaldehyde, 4-trimethylsilyl ethynyl, acmc-1bios, p-trimethylsilylethynylbenzaldehyde, 4-trimethylsilylethynylbenzaldehyde
PubChem CID 2771643
IUPAC Name 4-(2-trimethylsilylethynyl)benzaldehyde
SMILES C[Si](C)(C)C#CC1=CC=C(C=O)C=C1

Specifications

Melting Point 66°C to 70°C
Quantity 25 g
Formula Weight 202.33
Percent Purity 97%
Chemical Name or Material 4-[(Trimethylsilyl)ethynyl]benzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.