missing translation for 'onlineSavingsMsg'
Learn More

4-Trimethylsilyl-3-butyn-2-ol, 97%

Product Code. 11333696
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11333696

Brand: Thermo Scientific Alfa Aesar L14815.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6999-19-5 | C7H14OSi | 142.273 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6999-19-5
Molecular Formula C7H14OSi
Molecular Weight (g/mol) 142.273
MDL Number MFCD00190213
InChI Key HJJSDJHRTMFJLP-UHFFFAOYSA-N
Synonym 4-trimethylsilyl-3-butyn-2-ol, 4-trimethylsilyl but-3-yn-2-ol, 1-trimethylsilylbut-1-yne-3-ol, +/-4-trimethylsilyl-3-butyn-2-ol, acmc-20mpjc, 3-butyn-2-ol, 4-trimethylsilyl-, 2s, acmc-1bfey, hjjsdjhrtmfjlp-uhfffaoysa, 3-butyn-2-ol,4-trimethylsilyl
PubChem CID 2760828
IUPAC Name 4-trimethylsilylbut-3-yn-2-ol
SMILES CC(C#C[Si](C)(C)C)O

Specifications

Boiling Point 126°C to 129°C
Quantity 25 g
UN Number UN1987
Beilstein 1923632
Flash Point 57°C (135°F)
Formula Weight 142.27
Percent Purity 97%
Refractive Index 1.446
Density 0.846
Chemical Name or Material 4-Trimethylsilyl-3-butyn-2-ol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.